Gene Transcripts

Gene Symbol
Pxmp4
Entrez Gene ID
282634
Gene Name
peroxisomal membrane protein 4
Gene Aliases
Chromosome Location
Chr. 3: 150091646 - 150108893 on Build Rnor_6.0
UniGene ID
Rn.7752
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172223.2 NP_757377.1 4 590
GenBank mRNA AY151834.1 - 4 568
BC081699.1 - 4 590 7193564

View Full Product Details