Gene Transcripts

Gene Symbol
Yipf3
Entrez Gene ID
301245
Gene Name
Yip1 domain family, member 3
Gene Aliases
C6orf109, KLIP1, LRRGT00110, RGD1308185
Chromosome Location
Chr. 9: 17115308 - 17120618 on Build Rnor_6.0
UniGene ID
Rn.104143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007801.2 NP_001007802.1 2 257
GenBank mRNA AY387096.1 - 2 152
BC087102.1 - 2 257 7192818

View Full Product Details