Gene Transcripts

Gene Symbol
Rit1
Entrez Gene ID
499652
Gene Name
Ras-like without CAAX 1
Gene Aliases
RGD1559874
Chromosome Location
Chr. 2: 188087486 - 188099444 on Build Rnor_6.0
UniGene ID
Rn.50417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109185.1 NP_001102655.1 5 700

View Full Product Details