Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135776.1 | NP_001129248.1 | 2 | 1374 | |
NM_014007.3 | NP_054726.1 | 3 | 1497 | ||
XM_005251835.3 | XP_005251892.1 | 3 | 1818 | ||
XM_011518409.1 | XP_011516711.1 | 4 | 1510 | ||
XM_011518411.2 | XP_011516713.1 | 2 | 1624 | ||
GenBank mRNA | AB007874.1 | - | 1 | 2356 | |
AF049907.1 | - | 3 | 1477 | ||
AK026123.1 | - | 3 | 1443 | ||
BC008828.2 | - | 3 | 1421 | 4123095 | |
BT007194.1 | - | 1 | 1224 |