Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 7 | 1061 | |
NM_014456.4 | NP_055271.2 | 7 | 1103 | ||
NM_145341.3 | NP_663314.1 | 8 | 1187 | ||
GenBank mRNA | AK223623.1 | - | 7 | 977 | |
AK296753.1 | - | 7 | 928 | ||
AK312540.1 | - | 7 | 999 | ||
AK315295.1 | - | 8 | 1065 | ||
BC026104.2 | - | 7 | 970 | 4798293 | |
BC031049.1 | - | 7 | 970 | 5277621 | |
BX537500.1 | - | 6 | 4051 | ||
BX641168.1 | - | 3 | 2496 | ||
BX647834.1 | - | 7 | 945 | ||
U96628.1 | - | 7 | 901 |