Gene Transcripts

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
Chromosome Location
Chr. 13: 27919982 - 27926314 on Build GRCh38
UniGene ID
Hs.32938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000209.3 NP_000200.1 2 1287
GenBank mRNA U30329.1 - 2 1199
X99894.1 - 2 1278

View Full Product Details