Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006011.3 | NP_006002.1 | 1 | 234 | |
XM_017022643.1 | XP_016878132.1 | 1 | 233 | ||
GenBank mRNA | AK313924.1 | - | 1 | 182 | |
AW163487.1 | - | 234 | |||
BC069584.1 | - | 1 | 103 | ||
BC096202.3 | - | 1 | 103 | 40004644 | |
BC096203.3 | - | 1 | 103 | 40004645 | |
BC096204.3 | - | 1 | 103 | 40004646 | |
BC096205.3 | - | 1 | 103 | 40004648 | |
U33551.1 | - | 1 | 79 | ||
U82762.1 | - | 1 | 104 |