Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001244580.1 | NP_001231509.1 | 25 | 3668 | |
NM_003496.3 | NP_003487.1 | 25 | 3668 | ||
GenBank mRNA | AB209489.1 | - | 15 | 2603 | |
AF076974.1 | - | 25 | 3668 | ||
AF110377.1 | - | 25 | 3588 |