Gene Transcripts

Gene Symbol
PLD4
Entrez Gene ID
122618
Gene Name
phospholipase D family member 4
Gene Aliases
C14orf175
Chromosome Location
Chr. 14: 104924806 - 104937789 on Build GRCh38
UniGene ID
Hs.407101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308174.1 NP_001295103.1 4 630
NM_138790.3 NP_620145.2 4 592
XM_011536411.2 XP_011534713.1 4 640
XM_017020965.1 XP_016876454.1 4 529
GenBank mRNA AK295352.1 - 4 585
AK296188.1 - 4 630
AY358843.1 - 4 558
BC015003.1 - 4 521 4853026

View Full Product Details