Gene Transcripts
- Gene Symbol
- TAS2R43
- Entrez Gene ID
- 259289
- Gene Name
- taste 2 receptor member 43
- Gene Aliases
- T2R43, T2R52
- Chromosome Location
- Chr. 12: 11091287 - 11092313 on Build GRCh38
- UniGene ID
- Hs.688195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_176884.2 |
NP_795365.2
|
1 |
193 |
|
- Gene Symbol
- TAS2R46
- Entrez Gene ID
- 259292
- Gene Name
- taste 2 receptor member 46
- Gene Aliases
- T2R46, T2R54
- Chromosome Location
- Chr. 12: 11061365 - 11062294 on Build GRCh38
- UniGene ID
- Hs.688193
- Species
- Homo sapiens
View Full Product Details