Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr. 10: 32446082 - 32882864 on Build GRCh38
UniGene ID
Hs.585464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK023093.1 - 10 1257
AK097668.1 - 2 205
AK301824.1 - 7 647
BC125090.1 - 13 1426 40118176
BC125091.1 - 8 782 40118177

View Full Product Details