Gene Transcripts

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
Chromosome Location
Chr. 1: 147629658 - 147670543 on Build GRCh38
UniGene ID
Hs.562154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323625.1 NP_001310554.1 9 1576
NM_016361.4 NP_057445.4 9 1545
NR_136633.1 9 1531
NR_136634.1 10 1906
NR_136635.1 9 1531
NR_136636.1 9 1851
XM_011509601.2 XP_011507903.1 9 1501
XM_017001422.1 XP_016856911.1 9 1501
GenBank mRNA AB030039.2 - 9 1349
AB031478.1 - 9 1456
AB209248.1 - 8 3627
AK000657.1 - 9 1282
AY358552.1 - 9 1253
BC009965.1 - 9 1525 4299943
BC034686.1 - 9 1529 3463770
CR457101.1 - 9 1060
KJ534759.1 - 9 1081
KJ535036.1 - 8 953
KJ535044.1 - 6 719
KJ535055.1 - 9 1036

View Full Product Details