Gene Transcripts

Gene Symbol
FFAR3
Entrez Gene ID
2865
Gene Name
free fatty acid receptor 3
Gene Aliases
FFA3R, GPR41, GPR42
Chromosome Location
Chr. 19: 35358106 - 35360491 on Build GRCh38
UniGene ID
Hs.248055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005304.3 NP_005295.1 2 1620
XM_011526858.2 XP_011525160.1 2 1976
GenBank mRNA BC035657.1 - 2 1606

View Full Product Details