Gene Transcripts

Gene Symbol
SPG20
Entrez Gene ID
23111
Gene Name
spastic paraplegia 20 (Troyer syndrome)
Gene Aliases
SPARTIN, TAHCCP1
Chromosome Location
Chr. 13: 36301638 - 36370180 on Build GRCh38
UniGene ID
Hs.440414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142294.1 NP_001135766.1 2 486
NM_001142295.1 NP_001135767.1 2 610
NM_001142296.1 NP_001135768.1 3 662
NM_015087.4 NP_055902.1 2 679
XM_005266313.4 XP_005266370.1 2 604
XM_005266314.3 XP_005266371.1 3 653
XM_005266315.3 XP_005266372.1 3 507
XM_005266317.3 XP_005266374.1 2 437
XM_011535012.2 XP_011533314.1 2 590
GenBank mRNA AB011182.1 - 2 453
AK001949.1 - 2 443
AK291724.1 - 2 530
AY038359.1 - 1 392
AY123329.1 - 2 453
BC047083.1 - 2 486 5313378
BX648004.1 - 2 575
KU178563.1 - 1 392
KU178564.1 - 1 392

View Full Product Details