Gene Transcripts

Gene Symbol
Rsrp1
Entrez Gene ID
27981
Gene Name
arginine/serine rich protein 1
Gene Aliases
2700043I21Rik, D4Ucla2, D4Wsu53e
Chromosome Location
Chr. 4: 134923625 - 134927370 on Build GRCm38
UniGene ID
Mm.331964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023665.3 NP_076154.3 2, 3 762
GenBank mRNA AB041555.1 - 2, 3 742
AK003799.1 - 2, 3 763
AK012363.1 - 2, 3 763
AK028145.1 - 2, 3 764
AK029529.1 - 2, 3 762
AK150024.1 - 2, 3 763
AK150665.1 - 2, 3 742
AK152736.1 - 2, 3 742
AK160993.1 - 2, 3 763
AK169598.1 - 2, 3 764
AK169760.1 - 2, 3 763
AK178549.1 - 1, 2 108
AK206224.1 - 1, 2 108
BC043057.1 - 2, 3 747 6419597
BC046231.1 - 2, 3 664 6813388
BC054728.1 - 2, 3 735 6419723
BC056986.1 - 2, 3 696 6816649

View Full Product Details