Gene Transcripts
- Gene Symbol
- RGL1
- Entrez Gene ID
- 23179
- Gene Name
- ral guanine nucleotide dissociation stimulator like 1
- Gene Aliases
- RGL
- Chromosome Location
- Chr. 1: 183636047 - 183928551 on Build GRCh38
- UniGene ID
- Hs.497148
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297669.1 |
NP_001284598.1
|
12, 13 |
1910 |
|
NM_001297670.1 |
NP_001284599.1
|
11, 12 |
1800 |
|
NM_001297671.1 |
NP_001284600.1
|
11, 12 |
1536 |
|
NM_015149.4 |
NP_055964.3
|
12, 13 |
1911 |
|
XM_011509339.2 |
XP_011507641.1
|
13, 14 |
1957 |
|
XM_011509341.1 |
XP_011507643.1
|
12, 13 |
1864 |
|
XM_011509342.2 |
XP_011507644.1
|
12, 13 |
1864 |
|
XM_017000756.1 |
XP_016856245.1
|
11, 12 |
1385 |
|
GenBank mRNA |
AB023176.1 |
- |
11, 12 |
1476 |
|
AF186779.1 |
- |
11, 12 |
1497 |
|
AF186780.1 |
- |
12, 13 |
1873 |
|
AK295179.1 |
- |
11, 12 |
1674 |
|
AK295611.1 |
- |
8, 9 |
1113 |
|
AK316526.1 |
- |
12, 13 |
1848 |
|
AL080117.1 |
- |
10, 11 |
1279 |
|
BC136591.1 |
- |
12, 13 |
1615 |
9020581 |
View Full Product Details