Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011563.5 | NP_035693.3 | 4 | 464 | |
GenBank mRNA | AK008433.1 | - | 4 | 469 | |
AK010653.1 | - | 4 | 439 | ||
AK011963.1 | - | 411 | |||
AK014765.1 | - | 3 | 423 | ||
AK088280.1 | - | 4 | 483 | ||
BC002034.1 | - | 4 | 472 | 3485635 | |
BC081454.1 | - | 4 | 383 | 6818396 | |
BC086783.1 | - | 3 | 350 | 5700069 | |
U20611.1 | - | 4 | 426 | ||
U51679.1 | - | 4 | 401 | ||
X82067.1 | - | 4 | 433 |