Gene Transcripts

Gene Symbol
Ankrd11
Entrez Gene ID
77087
Gene Name
ankyrin repeat domain 11
Gene Aliases
2410104C19Rik, 3010027A04Rik, 6330578C09Rik, 9530048I21Rik, AA930108, Gm176, Yod
Chromosome Location
Chr. 8: 122883822 - 123042284 on Build GRCm38
UniGene ID
Mm.31567
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081379.2 NP_001074848.2 9 1708
GenBank mRNA AK144935.1 - 1 342
AK164512.1 - 6 4146
BC005576.1 - 7 1305
BC158043.1 - 8 1373 9088159
BC172141.1 - 9 1470 9054811

View Full Product Details