Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199252.2 | NP_954860.1 | 7 | 1049 | |
GenBank mRNA | AJ422143.1 | - | 7 | 1049 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142539.1 | NP_001136011.1 | 7 | 890 | |
GenBank mRNA | BC139460.1 | - | 7 | 890 | |
BC147830.1 | - | 7 | 890 | 9087547 | |
BC151195.1 | - | 7 | 890 | 9088083 |