Gene Transcripts

Gene Symbol
Paqr6
Entrez Gene ID
68957
Gene Name
progestin and adipoQ receptor family member VI
Gene Aliases
1500001B10Rik
Chromosome Location
Chr. 3: 88364072 - 88368541 on Build GRCm38
UniGene ID
Mm.441486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198410.3 NP_940802.1 8 1570
GenBank mRNA AK005086.1 - 2 809
AK139144.1 - 8 1571
BC030379.1 - 3 1195

View Full Product Details