Gene Transcripts

Gene Symbol
Vom2r72
Entrez Gene ID
689072
Gene Name
vomeronasal 2 receptor, 72
Gene Aliases
Chromosome Location
Chr. 14: 765993 - 822360 on Build Rnor_6.0
UniGene ID
Rn.217638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099517.1 NP_001092987.1 2463

View Full Product Details