Gene Transcripts

Gene Symbol
Urm1
Entrez Gene ID
311840
Gene Name
ubiquitin related modifier 1
Gene Aliases
RGD1306599
Chromosome Location
Chr. 3: 8389024 - 8405868 on Build Rnor_6.0
UniGene ID
Rn.6516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137562.2 NP_001131034.2 3 163
GenBank mRNA BC169094.1 - 3 153
CB313752.1 - 3 166

View Full Product Details