Gene Transcripts

Gene Symbol
Nudcd3
Entrez Gene ID
100125378
Gene Name
NudC domain containing 3
Gene Aliases
Chromosome Location
Chr. 14: 86326413 - 86333397 on Build Rnor_6.0
UniGene ID
Rn.217147
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103362.1 NP_001096832.1 549
GenBank mRNA BC107925.1 - 549

View Full Product Details