Gene Transcripts

Gene Symbol
Rsg1
Entrez Gene ID
500576
Gene Name
REM2 and RAB-like small GTPase 1
Gene Aliases
RGD1561421
Chromosome Location
Chr. 5: 159735008 - 159735956 on Build Rnor_6.0
UniGene ID
Rn.15685
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109272.1 NP_001102742.1 2 237

View Full Product Details