Gene Transcripts
- Gene Symbol
- CBX5
- Entrez Gene ID
- 23468
- Gene Name
- chromobox 5
- Gene Aliases
- HEL25, HP1, HP1A
- Chromosome Location
- Chr. 12: 54230946 - 54280131 on Build GRCh38
- UniGene ID
- Hs.349283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127321.1 |
NP_001120793.1
|
2, 3 |
266 |
|
NM_001127322.1 |
NP_001120794.1
|
2, 3 |
464 |
|
NM_012117.2 |
NP_036249.1
|
2, 3 |
312 |
|
GenBank mRNA |
AK313506.1 |
- |
2, 3 |
273 |
|
BC006821.1 |
- |
2, 3 |
214 |
3448801 |
BP236173.1 |
- |
2, 3 |
266 |
|
BP336150.1 |
- |
2, 3 |
312 |
|
CR457418.1 |
- |
1, 2 |
146 |
|
DT217255.1 |
- |
2, 3 |
464 |
|
EU794599.1 |
- |
2, 3 |
281 |
|
L07515.1 |
- |
2, 3 |
281 |
|
S62077.1 |
- |
2, 3 |
280 |
|
U26311.1 |
- |
1, 2 |
144 |
|
View Full Product Details