Gene Transcripts
- Gene Symbol
- SSBP2
- Entrez Gene ID
- 23635
- Gene Name
- single stranded DNA binding protein 2
- Gene Aliases
- HSPC116, SOSS-B2
- Chromosome Location
- Chr. 5: 81417360 - 81751253 on Build GRCh38
- UniGene ID
- Hs.102735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256732.1 |
NP_001243661.1
|
1, 2 |
274 |
|
NM_001256733.1 |
NP_001243662.1
|
1, 2 |
274 |
|
NM_001256734.1 |
NP_001243663.1
|
1, 2 |
274 |
|
NM_001256735.1 |
NP_001243664.1
|
1, 2 |
274 |
|
NM_001256736.1 |
NP_001243665.1
|
1, 2 |
274 |
|
NM_012446.3 |
NP_036578.2
|
1, 2 |
274 |
|
XM_011543297.2 |
XP_011541599.1
|
1, 2 |
189 |
|
XM_017009306.1 |
XP_016864795.1
|
1, 2 |
189 |
|
GenBank mRNA |
AF077048.1 |
- |
1, 2 |
111 |
|
AF161465.1 |
- |
1, 2 |
105 |
|
AK074537.1 |
- |
1, 2 |
69 |
|
AK293544.1 |
- |
1, 2 |
69 |
|
AK294864.1 |
- |
1, 2 |
69 |
|
AK297494.1 |
- |
1, 2 |
69 |
|
AK299858.1 |
- |
1, 2 |
118 |
|
AK312337.1 |
- |
1, 2 |
118 |
|
AL080076.1 |
- |
1, 2 |
122 |
|
BC017020.1 |
- |
1, 2 |
122 |
3913337 |
BI517313.1 |
- |
1, 2 |
274 |
|
View Full Product Details