Gene Transcripts

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
Chromosome Location
Chr. 20: 20052514 - 20360714 on Build GRCh38
UniGene ID
Hs.729168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015585.3 NP_056400.3 14 1599
XM_005260688.3 XP_005260745.1 14 1562
XM_005260689.2 XP_005260746.1 14 1562
XM_005260690.3 XP_005260747.1 13 1487
XM_005260691.3 XP_005260748.1 14 1562
XM_005260692.3 XP_005260749.1 14 1562
XM_005260693.3 XP_005260750.1 14 1562
XM_005260695.4 XP_005260752.1 9 1873
XM_005260696.4 XP_005260753.1 5 446
XM_005260697.1 XP_005260754.1 14 1562
XM_011529211.1 XP_011527513.1 13 1420
XM_011529212.2 XP_011527514.1 14 1562
XM_017027786.1 XP_016883275.1 14 1502
XM_017027787.1 XP_016883276.1 14 1502
XM_017027788.1 XP_016883277.1 14 1562
XM_017027789.1 XP_016883278.1 14 1562
XM_017027790.1 XP_016883279.1 14 1562
XM_017027791.1 XP_016883280.1 4 344
XM_017027792.1 XP_016883281.1 5 386
XM_017027793.1 XP_016883282.1 5 446
XM_017027794.1 XP_016883283.1 5 510
XM_017027795.1 XP_016883284.1 14 1562
GenBank mRNA AK304174.1 - 14 1502
BC028708.2 - 14 1561 4825941

View Full Product Details