Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025593.2 | NP_001020764.1 | 2 | 341 | |
NM_001025595.2 | NP_001020766.1 | 2 | 341 | ||
NM_001287431.1 | NP_001274360.1 | 2 | 206 | ||
NM_001287432.1 | NP_001274361.1 | 3 | 458 | ||
NM_001287433.1 | NP_001274362.1 | 3 | 323 | ||
NM_014447.3 | NP_055262.1 | 2 | 206 | ||
XM_011531869.2 | XP_011530171.1 | 3 | 355 | ||
XM_011531870.2 | XP_011530172.1 | 3 | 220 | ||
GenBank mRNA | AF124489.1 | - | 1 | 42 | |
AK096509.1 | - | 2 | 290 | ||
AK290472.1 | - | 2 | 177 | ||
AK299600.1 | - | 2 | 204 | ||
AK304141.1 | - | 2 | 206 | ||
AU099007.1 | - | 2 | 183 | ||
AU139844.1 | - | 3 | 198 | ||
BC103759.1 | - | 2 | 170 | 6176474 | |
BC105114.1 | - | 2 | 293 | 8144117 | |
BC105116.1 | - | 2 | 293 | 8144119 | |
BC143675.1 | - | 2 | 293 | 9052189 | |
BC143676.1 | - | 2 | 158 | 9052190 | |
CB127228.1 | - | 2 | 133 | ||
CN305403.1 | - | 2 | 289 | ||
DA452239.1 | - | 3 | 315 | ||
DC346238.1 | - | 2 | 312 | ||
U52521.1 | - | 2 | 172 |