Gene Transcripts

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
Chromosome Location
Chr. 13: 43023226 - 43109170 on Build GRCh38
UniGene ID
Hs.438830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013238.2 NP_037370.2 4 701
GenBank mRNA AF126743.1 - 5 723
AJ270691.1 - 1 67
AK311866.1 - 4 337
AY248898.1 - 4 701
AY512565.1 - 4 394
BC010910.1 - 4 430 4332619
BC095400.1 - 4 320 30530999

View Full Product Details