Gene Transcripts
- Gene Symbol
- SNX10
- Entrez Gene ID
- 29887
- Gene Name
- sorting nexin 10
- Gene Aliases
- OPTB8
- Chromosome Location
- Chr. 7: 26291895 - 26374330 on Build GRCh38
- UniGene ID
- Hs.741316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199835.1 |
NP_001186764.1
|
3, 4 |
369 |
|
NM_001199837.1 |
NP_001186766.1
|
1, 2 |
174 |
|
NM_001318198.1 |
NP_001305127.1
|
4, 5 |
443 |
|
NM_001318199.1 |
NP_001305128.1
|
3, 4 |
244 |
|
NM_013322.2 |
NP_037454.2
|
3, 4 |
322 |
|
XM_006715712.2 |
XP_006715775.1
|
3, 4 |
522 |
|
XM_017012085.1 |
XP_016867574.1
|
5, 6 |
537 |
|
XM_017012086.1 |
XP_016867575.1
|
5, 6 |
976 |
|
XM_017012087.1 |
XP_016867576.1
|
4, 5 |
466 |
|
XM_017012088.1 |
XP_016867577.1
|
4, 5 |
413 |
|
GenBank mRNA |
AF121860.1 |
- |
3, 4 |
235 |
|
AK296142.1 |
- |
4, 5 |
273 |
|
AK307905.1 |
- |
1, 2 |
123 |
|
AK312850.1 |
- |
3, 4 |
244 |
|
AK316088.1 |
- |
4, 5 |
419 |
|
BC031050.1 |
- |
3, 4 |
216 |
5278015 |
BC034992.1 |
- |
3, 4 |
295 |
4827599 |
BJ995940.1 |
- |
1, 2 |
174 |
|
BM147978.1 |
- |
3, 4 |
363 |
|
DA377368.1 |
- |
3, 4 |
244 |
|
View Full Product Details