Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317980.1 | NP_001304909.1 | 4 | 492 | |
NM_001317981.1 | NP_001304910.1 | 4 | 466 | ||
NM_001317982.1 | NP_001304911.1 | 4 | 448 | ||
NM_016093.3 | NP_057177.1 | 4 | 438 | ||
XM_005265921.3 | XP_005265978.1 | 4 | 489 | ||
XM_011534565.2 | XP_011532867.1 | 4 | 1053 | ||
XM_017009519.1 | XP_016865008.1 | 4 | 480 | ||
XM_017009520.1 | XP_016865009.1 | 4 | 411 | ||
GenBank mRNA | AF083248.1 | - | 4 | 412 | |
AK128872.1 | - | 1 | 406 | ||
BC017360.2 | - | 4 | 426 | 4551093 | |
BC070192.1 | - | 4 | 411 | 6727802 | |
BQ278240.1 | - | 4 | 439 | ||
BU193755.1 | - | 4 | 472 | ||
BU569345.1 | - | 4 | 451 |