Gene Transcripts
- Gene Symbol
- LUC7L2
- Entrez Gene ID
- 51631
- Gene Name
- LUC7 like 2, pre-mRNA splicing factor
- Gene Aliases
- CGI-59, CGI-74, LUC7B2
- Chromosome Location
- Chr. 7: 139340359 - 139423457 on Build GRCh38
- UniGene ID
- Hs.731488
- Species
- Homo sapiens
- Gene Symbol
- C7orf55-LUC7L2
- Entrez Gene ID
- 100996928
- Gene Name
- C7orf55-LUC7L2 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 7: 139341132 - 139423457 on Build GRCh38
- UniGene ID
- Hs.731488
- Species
- Homo sapiens
- Gene Symbol
- LOC100129148
- Entrez Gene ID
- 100129148
- Gene Name
- uncharacterized LOC100129148
- Gene Aliases
- Chromosome Location
- Chr. 7: 139417462 - 139427526 on Build GRCh38
- UniGene ID
- Hs.744449
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI811258.1 |
- |
1 |
131 |
|
View Full Product Details