Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001272037.1 | NP_001258966.1 | 8 | 1088 | |
NM_144991.2 | NP_659428.2 | 7 | 1026 | ||
GenBank mRNA | AJ487962.1 | - | 7 | 1026 | |
BC021197.2 | - | 7 | 915 | ||
BC115415.1 | - | 5 | 552 | 40080529 |