Gene Transcripts
- Gene Symbol
- ACER3
- Entrez Gene ID
- 55331
- Gene Name
- alkaline ceramidase 3
- Gene Aliases
- APHC, PHCA
- Chromosome Location
- Chr. 11: 76860873 - 77026797 on Build GRCh38
- UniGene ID
- Hs.23862
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300953.1 |
NP_001287882.1
|
2, 3 |
268 |
|
NM_001300954.1 |
NP_001287883.1
|
3, 4 |
450 |
|
NM_001300955.1 |
NP_001287884.1
|
2, 3 |
326 |
|
NM_018367.6 |
NP_060837.3
|
3, 4 |
379 |
|
XM_011545151.2 |
XP_011543453.1
|
3, 4 |
345 |
|
XM_011545152.2 |
XP_011543454.1
|
3, 4 |
315 |
|
XM_011545153.2 |
XP_011543455.1
|
3, 4 |
345 |
|
XM_017017987.1 |
XP_016873476.1
|
3, 4 |
364 |
|
XM_017017988.1 |
XP_016873477.1
|
3, 4 |
345 |
|
GenBank mRNA |
AF214454.1 |
- |
3, 4 |
333 |
|
AF327353.1 |
- |
3, 4 |
344 |
|
AK002100.1 |
- |
4, 5 |
439 |
|
AK293800.1 |
- |
4, 5 |
350 |
|
AK294978.1 |
- |
2, 3 |
199 |
|
AK295142.1 |
- |
3, 4 |
367 |
|
AK295327.1 |
- |
4, 5 |
463 |
|
AK308856.1 |
- |
4, 5 |
436 |
|
AK315000.1 |
- |
3, 4 |
345 |
|
AK316120.1 |
- |
2, 3 |
264 |
|
BC063034.1 |
- |
4, 5 |
339 |
|
BC073853.1 |
- |
3, 4 |
296 |
3867010 |
BG702017.1 |
- |
3, 4 |
384 |
|
DB461413.1 |
- |
3, 4 |
370 |
|
View Full Product Details