Gene Transcripts

Gene Symbol
NAGK
Entrez Gene ID
55577
Gene Name
N-acetylglucosamine kinase
Gene Aliases
GNK, HSA242910
Chromosome Location
Chr. 2: 71068246 - 71078868 on Build GRCh38
UniGene ID
Hs.7036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017567.4 NP_060037.3 7 1000
XM_005264412.4 XP_005264469.1 7 656
XM_005264413.1 XP_005264470.1 7 757
XM_005264415.2 XP_005264472.1 6 868
XM_011532957.1 XP_011531259.1 7 965
GenBank mRNA AJ242910.1 - 7 611
AK001812.1 - 7 629
AK092876.1 - 1 950
AK097223.1 - 6 2418
AK172854.1 - 1 321
AK222645.1 - 7 628
AK297224.1 - 7 977
AL832837.1 - 3 3840
BC001029.2 - 7 642 3347484
BC005371.1 - 7 632 3932248
CR457271.1 - 7 594

View Full Product Details