Gene Transcripts

Gene Symbol
ZNF280C
Entrez Gene ID
55609
Gene Name
zinc finger protein 280C
Gene Aliases
SUHW3, ZNF633
Chromosome Location
Chr. X: 130202699 - 130268948 on Build GRCh38
UniGene ID
Hs.308418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017666.4 NP_060136.1 8 902
XM_006724765.3 XP_006724828.1 8 853
GenBank mRNA AK000102.1 - 8 825
AK290373.1 - 8 853
AL133204.1 - 8 837
AL834333.1 - 8 833
BC051728.1 - 8 853 4828435

View Full Product Details