Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 5 | 544 | |
XM_005256045.3 | XP_005256102.1 | 4 | 429 | ||
XM_011523227.2 | XP_011521529.1 | 5 | 1274 | ||
GenBank mRNA | AF175265.1 | - | 5 | 502 | |
AF183418.1 | - | 4 | 453 | ||
AF186382.1 | - | 5 | 498 | ||
AF191298.2 | - | 5 | 472 | ||
AK001614.1 | - | 5 | 492 | ||
AK023650.1 | - | 1 | 55 | ||
AK223218.1 | - | 5 | 463 | ||
AL136888.1 | - | 5 | 492 | ||
BC002414.2 | - | 4 | 451 | 3162255 | |
BC010362.1 | - | 5 | 465 | 4249949 | |
BC093036.1 | - | 5 | 490 | 30340379 | |
DB049686.1 | - | 5 | 544 |