Gene Transcripts

Gene Symbol
PCDHB15
Entrez Gene ID
56121
Gene Name
protocadherin beta 15
Gene Aliases
PCDH-BETA15
Chromosome Location
Chr. 5: 141245349 - 141248234 on Build GRCh38
UniGene ID
Hs.130757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018935.3 NP_061758.1 1 1103
GenBank mRNA AF152494.1 - 1 873
AF217742.1 - 1 873
AK312794.1 - 1 1105
BC038797.1 - 1 1039 5767743

View Full Product Details