Gene Transcripts

Gene Symbol
RETN
Entrez Gene ID
56729
Gene Name
resistin
Gene Aliases
ADSF, FIZZ3, RETN1, RSTN, XCP1
Chromosome Location
Chr. 19: 7669086 - 7670454 on Build GRCh38
UniGene ID
Hs.283091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193374.1 NP_001180303.1 2 105
NM_020415.3 NP_065148.1 2 114
GenBank mRNA AB111910.1 - 1 68
AF205952.1 - 1 92
AF290874.1 - 2 114
AF323081.1 - 2 114
AY207314.1 - 2 105
AY359066.1 - 1 92
BC069302.1 - 2 114 7262685
BC101554.1 - 2 97 8069060
BC101560.1 - 2 97 8069066

View Full Product Details