Gene Transcripts

Gene Symbol
NT5M
Entrez Gene ID
56953
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
dNT-2, dNT2, mdN
Chromosome Location
Chr. 17: 17303366 - 17347663 on Build GRCh38
UniGene ID
Hs.513977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020201.3 NP_064586.1 4 666
XM_005256731.3 XP_005256788.1 4 655
XM_005256732.3 XP_005256789.1 4 655
XM_005256733.3 XP_005256790.1 4 655
XM_005256734.3 XP_005256791.1 4 655
XM_005256737.1 XP_005256794.1 5 468
XM_011523963.1 XP_011522265.1 6 505
XM_011523964.2 XP_011522266.1 6 474
XM_011523965.2 XP_011522267.1 5 378
XM_011523967.2 XP_011522269.1 5 748
XM_017024879.1 XP_016880368.1 5 1408
GenBank mRNA AF210652.1 - 4 697
BC035838.1 - 4 640 5727827
DQ323995.1 - 4 481

View Full Product Details