Gene Transcripts

Gene Symbol
YAE1D1
Entrez Gene ID
57002
Gene Name
Yae1 domain containing 1
Gene Aliases
C7orf36, GK003
Chromosome Location
Chr. 7: 39566376 - 39612089 on Build GRCh38
UniGene ID
Hs.72168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020192.4 NP_064577.1 3 582
GenBank mRNA AF226046.1 - 3 548
AK314937.1 - 3 554
BC022043.1 - 3 552 4734433
BM767075.1 - 3 582
BX648555.1 - 1 3135
CR457198.1 - 3 539

View Full Product Details