Gene Transcripts

Gene Symbol
RALGAPB
Entrez Gene ID
57148
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
Gene Aliases
KIAA1219, RalGAPbeta
Chromosome Location
Chr. 20: 38472806 - 38578861 on Build GRCh38
UniGene ID
Hs.598850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282917.1 NP_001269846.1 19 2986
NM_001282918.1 NP_001269847.1 19 3116
NM_020336.3 NP_065069.1 19 3128
XM_005260462.2 XP_005260519.1 20 3122
XM_005260463.2 XP_005260520.1 20 3122
XM_005260464.2 XP_005260521.1 20 3110
XM_005260465.2 XP_005260522.1 19 3086
XM_017027966.1 XP_016883455.1 20 3110
XM_017027967.1 XP_016883456.1 19 3074
XM_017027968.1 XP_016883457.1 19 2648
XM_017027969.1 XP_016883458.1 20 3139
XM_017027970.1 XP_016883459.1 4 610
GenBank mRNA AB033045.3 - 19 3106
AK125260.1 - 7 889
AK125916.1 - 14 2085
AL834436.1 - 17 2350
BC047482.1 - 19 3091 5270777
CR936800.1 - 19 2892

View Full Product Details