Gene Transcripts

Gene Symbol
RAB40C
Entrez Gene ID
57799
Gene Name
RAB40C, member RAS oncogene family
Gene Aliases
RARL, RASL8C
Chromosome Location
Chr. 16: 589357 - 629273 on Build GRCh38
UniGene ID
Hs.459630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172663.1 NP_001166134.1 5 518
NM_001172664.1 NP_001166135.1 5 410
NM_001172665.1 NP_001166136.1 5 370
NM_001172666.1 NP_001166137.1 5 370
NM_021168.4 NP_066991.3 4 415
GenBank mRNA AY823398.1 - 4 340
BC028696.1 - 4 417 4826800
BG696681.1 - 5 519
BG715678.1 - 5 412
BM545442.1 - 4 489
BQ685517.1 - 5 349
BX442451.2 - 5 459
CX783728.1 - 5 503

View Full Product Details