Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165960.1 | NP_001159432.1 | 6 | 1163 | |
NM_021628.2 | NP_067641.2 | 6 | 999 | ||
XM_017024921.1 | XP_016880410.1 | 5 | 942 | ||
XM_017024922.1 | XP_016880411.1 | 5 | 942 | ||
XM_017024923.1 | XP_016880412.1 | 5 | 942 | ||
XM_017024924.1 | XP_016880413.1 | 5 | 942 | ||
XM_017024925.1 | XP_016880414.1 | 5 | 942 | ||
GenBank mRNA | AF182218.1 | - | 5 | 830 | |
AJ269499.1 | - | 5 | 1128 | ||
AK292732.1 | - | 5 | 902 | ||
AK296416.1 | - | 6 | 1064 | ||
AK299742.1 | - | 6 | 1106 | ||
AK313677.1 | - | 6 | 963 | ||
BC101938.1 | - | 5 | 851 | 40011855 | |
BC101939.1 | - | 5 | 851 | 40011856 | |
BC103508.1 | - | 5 | 851 | 40011857 | |
BC104724.1 | - | 5 | 851 | 40011849 |