Gene Transcripts

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
Chromosome Location
Chr. 9: 136363956 - 136373681 on Build GRCh38
UniGene ID
Hs.694071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052813.4 NP_434700.2 2 336
NM_052814.3 NP_434701.1 2 336
GenBank mRNA AF311287.1 - 2 297
AK024001.1 - 2 316
AK292081.1 - 2 288
AK295647.1 - 2 175
AK309043.1 - 2 301
BC008877.2 - 2 311 4139529
BI524042.1 - 2 321
BM148244.1 - 2 336

View Full Product Details