Gene Transcripts

Gene Symbol
OSBPL10
Entrez Gene ID
114884
Gene Name
oxysterol binding protein like 10
Gene Aliases
ORP10, OSBP9
Chromosome Location
Chr. 3: 31660825 - 31981850 on Build GRCh38
UniGene ID
Hs.150122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174060.1 NP_001167531.1 6 1666
NM_017784.4 NP_060254.2 7 1858
XM_005264843.3 XP_005264900.1 7 1634
XM_005264844.4 XP_005264901.1 6 1442
XM_005264845.1 XP_005264902.1 2 542
XM_011533326.2 XP_011531628.1 7 1011
XM_017005667.1 XP_016861156.1 7 1524
XM_017005668.1 XP_016861157.1 7 1501
XM_017005669.1 XP_016861158.1 7 1030
XM_017005670.1 XP_016861159.1 4 1713
GenBank mRNA AB209872.1 - 4 493
AF392451.1 - 8 1568
AK000370.1 - 5 734
AK304067.1 - 6 1119
AL833247.1 - 7 961
BC003168.1 - 7 1262 2905981

View Full Product Details