Gene Transcripts

Gene Symbol
RIC3
Entrez Gene ID
79608
Gene Name
RIC3 acetylcholine receptor chaperone
Gene Aliases
AYST720, PRO1385
Chromosome Location
Chr. 11: 8092965 - 8169088 on Build GRCh38
UniGene ID
Hs.231850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135109.2 NP_001128581.1 2 260
NM_001206671.2 NP_001193600.1 6 806
NM_001206672.2 NP_001193601.1 4 563
NM_024557.4 NP_078833.3 6 803
NR_045405.1 7 855
XM_006718317.3 XP_006718380.1 6 935
XM_006718318.3 XP_006718381.1 6 848
XM_017018287.1 XP_016873776.1 6 935
XM_017018288.1 XP_016873777.1 7 932
XM_017018289.1 XP_016873778.1 7 879
XM_017018290.1 XP_016873779.1 5 702
GenBank mRNA AK021670.1 - 5 827
AK293933.1 - 6 853
AK296505.1 - 7 820
AK315379.1 - 6 762
AL832601.2 - 2 221
AM422214.1 - 4 509
AY326435.1 - 6 752
BC022455.1 - 6 785 4792930
BX647514.1 - 3 403
DA186330.1 - 2 260
KU178773.1 - 6 749
KU178774.1 - 5 655
KU178775.1 - 2 206

View Full Product Details