Gene Transcripts

Gene Symbol
ASB13
Entrez Gene ID
79754
Gene Name
ankyrin repeat and SOCS box containing 13
Gene Aliases
Chromosome Location
Chr. 10: 5638857 - 5666595 on Build GRCh38
UniGene ID
Hs.445899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024701.3 NP_078977.2 5 742
NR_024581.1 4 591
NR_037164.1 6 823
GenBank mRNA AF403031.1 - 5 724
AK023196.1 - 5 725
AK091935.1 - 4 591
AK292071.1 - 5 728
BC012056.1 - 6 800 4552643
CR457302.1 - 5 698

View Full Product Details