Gene Transcripts
- Gene Symbol
- NAA25
- Entrez Gene ID
- 80018
- Gene Name
- N(alpha)-acetyltransferase 25, NatB auxiliary subunit
- Gene Aliases
- C12orf30, MDM20, NAP1
- Chromosome Location
- Chr. 12: 112026689 - 112109022 on Build GRCh38
- UniGene ID
- Hs.530941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024953.3 |
NP_079229.2
|
3, 4 |
347 |
|
XM_006719606.2 |
XP_006719669.1
|
3, 4 |
1007 |
|
XM_017019977.1 |
XP_016875466.1
|
1, 2 |
70 |
|
GenBank mRNA |
AB054990.1 |
- |
3, 4 |
289 |
|
AK292485.1 |
- |
3, 4 |
320 |
|
AK307494.1 |
- |
4, 5 |
496 |
|
AV760516.2 |
- |
3, 4 |
312 |
|
BC034357.1 |
- |
3, 4 |
301 |
|
BC065481.1 |
- |
3, 4 |
280 |
|
BC113585.1 |
- |
3, 4 |
347 |
8322637 |
BC113587.1 |
- |
3, 4 |
347 |
8322639 |
BX641140.1 |
- |
3, 4 |
302 |
|
BX647450.1 |
- |
3, 4 |
300 |
|
View Full Product Details