Gene Transcripts

Gene Symbol
NRSN2
Entrez Gene ID
80023
Gene Name
neurensin 2
Gene Aliases
C20orf98, dJ1103G7.6
Chromosome Location
Chr. 20: 346726 - 359705 on Build GRCh38
UniGene ID
Hs.416024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323679.1 NP_001310608.1 6 989
NM_001323680.1 NP_001310609.1 4 1070
NM_001323681.1 NP_001310610.1 4 1061
NM_001323682.1 NP_001310611.1 5 1127
NM_001323683.1 NP_001310612.1 5 1121
NM_001323684.1 NP_001310613.1 5 1149
NM_001323685.1 NP_001310614.1 4 790
NM_024958.2 NP_079234.1 4 1048
NR_136649.1 3 853
XM_006723630.3 XP_006723693.1 4 1471
XM_011529360.1 XP_011527662.1 4 754
XM_011529362.2 XP_011527664.1 5 761
XM_011529363.2 XP_011527665.1 4 739
XM_017028073.1 XP_016883562.1 4 693
XM_017028074.1 XP_016883563.1 4 747
XM_017028075.1 XP_016883564.1 4 919
GenBank mRNA AK026982.1 - 4 661
AK290527.1 - 4 1025
AL136915.1 - 3 643
BC001963.1 - 3 628 3454427
BE792891.1 - 2 412
CR533441.1 - 2 510

View Full Product Details